Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRCP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552403
Description
Immunogen sequence: TPCRHQSPEI VREFLTALKS HKLTKAEKLQ LLNHRPVTAV EIQLMVEESE ERLTEEQIEA LLHTVTSILP AEPEAEQKKN TNSNVAMDEE DPA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.
This gene encodes a membrane protein that functions as part of a receptor complex for a small neuropeptide that increases intracellular cAMP levels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
CRCP | |
Polyclonal | |
Unconjugated | |
CRCP | |
AL022669; calcitonin gene-related peptide-receptor component protein; CGRP receptor component; CGRPRCP; CGRP-RCP; CGRP-receptor component protein; Crcp; DNA-directed RNA polymerase III subunit RPC9; RCP; RCP9; RNA polymerase III subunit C9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27297 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75575 | |
CRCP | |
Recombinant protein corresponding to Human CRCP. Recombinant protein control fragment (Product #RP-88678). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction