Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRCP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592273
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592273 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592273 Supplier Invitrogen™ Supplier No. PA592273
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEVKDANSAL LSNYETLKYI SKTPCRHQSP EIVREFLTAL KSHKLTKAEK LQLLNHRPVT AVEIQLMVEE SEERLTEEQI EALLHTVTSI LPAEPEAEQK KNTNSNVAMD EEDPA; Positive Samples: U-87MG, HeLa, Jurkat, Mouse testis; Cellular Location: Cell membrane, Cytoplasmic side, Nucleus, Peripheral membrane protein.

This gene encodes a membrane protein that functions as part of a receptor complex for a small neuropeptide that increases intracellular cAMP levels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRCP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CRCP
Gene Accession No. O35427, O75575, Q8VHM6
Gene Alias AL022669; calcitonin gene-related peptide-receptor component protein; CGRP receptor component; CGRPRCP; CGRP-RCP; CGRP-receptor component protein; Crcp; DNA-directed RNA polymerase III subunit RPC9; RCP; RCP9; RNA polymerase III subunit C9
Gene Symbols CRCP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-115 of human CRCP (NP_0010357371).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114205, 12909, 27297
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.