Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRCP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103800
Description
Antibody detects endogenous levels of total RPC9.
This gene encodes a membrane protein that functions as part of a receptor complex for a small neuropeptide that increases intracellular cAMP levels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
CRCP | |
Polyclonal | |
Unconjugated | |
CRCP | |
AL022669; calcitonin gene-related peptide-receptor component protein; CGRP receptor component; CGRPRCP; CGRP-RCP; CGRP-receptor component protein; Crcp; DNA-directed RNA polymerase III subunit RPC9; RCP; RCP9; RNA polymerase III subunit C9 | |
Rabbit | |
Affinity chromatography | |
RUO | |
27297 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O75575 | |
CRCP | |
A synthesized peptide derived from human CRCP(Accession O75575), corresponding to amino acid residues K4-S54. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction