Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598839
Description
CRD1 Polyclonal Antibody for Western Blot, ELISA
CRD1 catalyzes the formation of the isocyclic ring in chlorophyll biosynthesis. It mediates the cyclase reaction, which results in the formation of divinylprotochlorophyllide (Pchlide) characteristic of all chlorophylls from magnesium-protoporphyrin IX 13-monomethyl ester (MgPMME).
Specifications
| CRD1 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Protein G | |
| RUO | |
| Plant | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 3.63 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Recombinant Arabidopsis thaliana Magnesium-protoporphyrin IX monomethyl ester [oxidative] cyclase, chloroplastic protein (37-409AA). | |
| 100 μL | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction