Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cre Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056133G
Description
Cre Polyclonal specifically detects Cre in Mouse, Cre/lox System samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Cre | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Cyclization recombinase | |
A synthetic peptide made to a portion of bacteriophage protein CRE (within amino acids 50-100). [UniProt# P06956] | |
0.05 mL | |
Primary | |
Cre/lox Systems and Mouse. Not yet tested in other species. | |
Store at 4C in the dark. | |
Cre/lox Systems |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rabbit | |
Affinity Purified | |
RUO | |
2777477 | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction