Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cre Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056133F
Description
Cre Polyclonal specifically detects Cre in Mouse, Cre/lox System samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Cre | |
Polyclonal | |
Immunohistochemistry 1:4000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen 1:1000-1:5000 | |
Cyclization recombinase | |
A synthetic peptide made to a portion of bacteriophage protein CRE (within amino acids 50-100). [UniProt# P06956] | |
0.05 mL | |
Primary | |
Cre/lox Systems and Mouse. Not yet tested in other species. | |
Store at 4C in the dark. | |
Cre/lox Systems |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
FITC | |
PBS with 0.05% Sodium Azide | |
Rabbit | |
Affinity Purified | |
RUO | |
2777477 | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction