Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cre, Rabbit, Unlabeled, Polyclonal,

Catalog No. 690503MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
69-050-3MI 50 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-050-3MI Supplier MilliporeSigma™ Supplier No. 690503

Rabbit Polyclonal Antibody

The Anti-Cre Antibody is a rabbit polyclonal antibody that has high specificity and affinity for bacteriophage P1 Cre recombinase. This preparation can be used to monitor the presence of Cre protein in bacterial and mammalian systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cre
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Stabilized solution of purified IgG in 50% glycerol
Host Species Rabbit
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific, sensitive detection of Cre recombinase in cells and cell lysates
Content And Storage -20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.