Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cre recombinase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532245 Supplier Invitrogen™ Supplier No. PA532245
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T transfected recombinant cre protein, IPTG-induced cell lysate from E. coli expressing Cre recombinantproteins. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cre-recombinase is a 38 kDa DNA recombinase derived from the P1 bacteriophage. It is highly specific for a 34 bp DNA sequence (loxP) found in P1 DNA. It catalyzes site-specific recombination between two 34-base-pair LOXP sites. Its role is to maintain the phage genome as a monomeric unit-copy plasmid in the lysogenic state. It is homotetramer when bound to DNA and belongs to the phage integrase family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cre recombinase
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Alias Cre
Host Species Rabbit
Immunogen Recombinant protein conjugated synthetic peptide encompassing a sequence within the center region of Cre recombinase. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.