Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatine, anhydrous, 98%

Catalog No. aaa1747714
Click to view available options
Quantity:
25 g
100 g

Catalog No. AAA1747714

Supplier: Thermo Scientific Chemicals A1747714

Only null left
Add to Cart

CAS: 57-00-1 | C4H9N3O2 | 131.135 g/mol

Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-00-1
Molecular Formula C4H9N3O2
Molecular Weight (g/mol) 131.135
MDL Number MFCD00004282
InChI Key CVSVTCORWBXHQV-UHFFFAOYSA-N
Synonym creatine, creatin, kreatin, krebiozon, n-amidinosarcosine, n-methyl-n-guanylglycine, methylglycocyamine, 2-1-methylguanidino acetic acid, creatine, hydrate, alpha-methylguanido acetic acid
PubChem CID 586
ChEBI CHEBI:16919
IUPAC Name 2-[carbamimidoyl(methyl)amino]acetic acid
SMILES CN(CC(=O)O)C(=N)N

Specifications

Melting Point ∼290°C (decomposition)
Density 1.33
Assay Percent Range 98%
Quantity 25 g
Beilstein 907175
Merck Index 14,2568
Solubility Information Soluble in water.
Formula Weight 131.13
Percent Purity 98%
Chemical Name or Material Creatine, anhydrous
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 200-306-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.