Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Creatine Kinase BB Antibody (2ba6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259601H
Description
Creatine Kinase BB Monoclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
Creatine Kinase BB | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
2ba6 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
Creatine Kinase Isoenzyme | |
Human Creatine Kinase BB protein | |
0.1 mL | |
Cancer | |
1152 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction