Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Creatine Kinase BB Antibody (8E4), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259462APC
This item is not returnable.
View return policy
Description
Creatine Kinase BB Monoclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for Flow CytometrySpecifications
Creatine Kinase BB | |
Monoclonal | |
APC | |
Creatine Kinase Isoenzyme | |
Recombinant human Creatine Kinase BB (1-381aa) purified from E. coli | |
0.1 mL | |
Cancer | |
1152 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry | |
80000 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction