Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Creatine Kinase BB Antibody (CPTC-CKB-2), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308866AF350
Description
Creatine Kinase BB Monoclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Creatine Kinase BB | |
Monoclonal | |
Alexa Fluor 350 | |
Creatine Kinase Isoenzyme | |
Recombinant human full-length Creatine Kinase BB protein (Uniprot: P12277) | |
100 μg | |
Cancer | |
1152 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, Immunohistochemistry (Paraffin) | |
CPTC-CKB-2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction