Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Creatine Kinase BB Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300288JF646
Description
Creatine Kinase BB Polyclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
Creatine Kinase BB | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
Creatine Kinase Isoenzyme | |
Produced in rabbits immunized with purified, recombinant Human Creatine Kinase BB (Accession#: P12277; Pro2-Lys381) | |
0.1 mL | |
Cancer | |
1152 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction