Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Creatine Kinase BB Monoclonal Antibody (CK-CYK/21E10)
GREENER_CHOICE

Catalog No. PIMA533339
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533339 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533339 Supplier Invitrogen™ Supplier No. MA533339
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Product is supplied at 1 mL (100 μL/mL). Application notes: Western blot - Under reducing conditions, antibody detected a protein with a band size of 43-46 kDa. IHC-F- tissue was fixed for 2 hours in 2% paraformaldehyde and 0.1 % glutaraldehyde in 0.1 M PB. The tissue was stored until use in 1 % paraformaldehyde. The tissue was blocked with 1% normal swine serum. IHC-P - antigen retrieval: 0.5 % pepsin in 0.01 N HCl for 15 min at 37 °C; endogenous peroxidise blocking by using 3 % H2O2 in PBS for 30 min; blocking with 1% normal swine serum for 30 min.; Purkinje cells serve as positive control and connective tissue as negative control. Positive control: Brain lysate, Human Purknije cells; Negative control: Human fibroblasts.

The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Creatine Kinase BB
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CK-CYK/21E10
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene CKB
Gene Accession No. P00567, P07335, P12277, Q04447
Gene Alias Bck; B-CK; brain creatine kinase; Ck3; Ck-3; CKB; Ckbb; Ckbr; CPK-B; creatine kinase B; creatine kinase B chain; creatine kinase brain; creatine kinase brain-type; Creatine kinase B-type; creatine kinase, brain; creatine kinase-B; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; HEL-211; HEL-S-29
Gene Symbols CKB
Host Species Mouse
Immunogen Synthetic 17-mer peptide corresponding to the N-terminal sequence of human creatine kinase brain-type (SNSHNALKLRFPAEDEF).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009085, 1152, 12709, 24264
Target Species Human, Mouse, Rat, Rabbit
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.