Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Creatine Kinase MB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580478
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CKM/CK-MM.
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
Specifications
Creatine Kinase MB | |
Polyclonal | |
Unconjugated | |
Ckb | |
Bck; B-CK; Brain creatine kinase; CK MB; Ck3; Ck-3; CKB; Ckbb; Ckbr; Ckm; CKMM; CPK-B; CPK-M; creatine kinase B; creatine kinase B chain; creatine kinase brain; creatine kinase brain-type; Creatine kinase B-type; creatine kinase M chain; Creatine kinase M-type; creatine kinase, brain; creatine kinase, M-type; creatine kinase, muscle; creatine kinase-B; creatine kinase-M; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; HEL-211; HEL-S-29; MCK; M-CK | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
1152, 1158 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with no preservative | |
P06732, P12277 | |
Ckb, CKM | |
Recombinant Human CKM/CK-MM Protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction