Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Creatine kinase MT 1A Antibody (17A2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242664H
Description
Creatine kinase MT 1A Monoclonal specifically detects Creatine kinase MT 1A in Human samples. It is validated for Western Blot, ELISA.Specifications
Creatine kinase MT 1A | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CKMT1A | |
Recombinant human CKMT1A (40-417aa) purified from E. coli | |
100 μL | |
Protein Kinase | |
548596 | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, ELISA | |
17A2 | |
Western Blot, ELISA | |
Acidic-Type Mitochondrial Creatine Kinase, CKMT, CKMT1, CKMT1A, Creatine Kinase U-Type, Mitochondrial, Creatine Kinase, Mitochondrial 1 (Ubiquitous), Creatine Kinase, Mitochondrial 1A, EC 2.7.3.2, Mia-CK, Ubiquitous Mitochondrial Creatine Kinase, U-MtCK | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction