Learn More
Invitrogen™ Creatine Kinase MT Polyclonal Antibody

Description
Recommended positive controls: 293T, H1299, HeLaS3, HepG2. Predicted reactivity: Rat (98%), Xenopus laevis (88%), Rabbit (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Creatine Kinase MT |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | CKMT1A |
| Gene Accession No. | P12532 |
| Gene Alias | acidic-type mitochondrial creatine kinase; CKMT; CKMT1; CKMT1A; CKMT1B; Creatine kinase U-type, mitochondrial; creatine kinase, mitochondrial 1 (ubiquitous); creatine kinase, mitochondrial 1, ubiquitous; creatine kinase, mitochondrial 1A; creatine kinase, mitochondrial 1B; mia-CK; mi-CK; Mt-CK; ScCKmit; UbCKmit; ubiquitous mitochondrial creatine kinase; UMTCK; U-MtCK |
| Gene Symbols | CKMT1A, CKMT1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.