Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CKMT1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560387
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560387 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560387 Supplier Invitrogen™ Supplier No. PA560387
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YLIDCERRLE RGQDIRIPTP VIHTKH Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 81%.

Mitochondrial creatine kinase (MtCK) is responsible for the transfer of high energy phosphate from mitochondria to the cytosolic carrier, creatine. It belongs to the creatine kinase isoenzyme family. It exists as two isoenzymes, sarcomeric MtCK and ubiquitous MtCK, encoded by separate genes. Mitochondrial creatine kinase occurs in two different oligomeric forms: dimers and octamers, in contrast to the exclusively dimeric cytosolic creatine kinase isoenzymes. Many malignant cancers with poor prognosis have shown overexpression of ubiquitous mitochondrial creatine kinase, this may be related to high energy turnover and failure to eliminate cancer cells via apoptosis. Ubiquitous mitochondrial creatine kinase has 80% homology with the coding exons of sarcomeric mitochondrial creatine kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKMT1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CKMT1A
Gene Accession No. P12532
Gene Alias Acidic-type mitochondrial creatine kinase; CKMT; Ckmt1; CKMT1A; Ckmt1b; Creatine kinase U-type, mitochondrial; creatine kinase, mitochondrial 1 (ubiquitous); creatine kinase, mitochondrial 1, ubiquitous; creatine kinase, mitochondrial 1A; creatine kinase, mitochondrial 1B; mia-CK; mi-CK; Mt-CK; ScCKmit; UbCKmit; Ubiquitous mitochondrial creatine kinase; U-MtCK
Gene Symbols CKMT1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CKMT1A. Recombinant protein control fragment (Product #RP-99584).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 548596
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.