Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Creatine Kinase, Muscle/CKMM Antibody (CKMM/3341), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308304UV
Description
Creatine Kinase, Muscle/CKMM Monoclonal antibody specifically detects Creatine Kinase, Muscle/CKMM in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein ArraySpecifications
Creatine Kinase, Muscle/CKMM | |
Monoclonal | |
DyLight 350 | |
CKMMcreatine kinase M-type, Creatine Kinase Isoenzyme, Creatine kinase M chain, creatine kinase, muscle, creatine kinase-M, EC 2.7.3, EC 2.7.3.2, M-CK | |
Recombinant full-length Creatine Kinase, Muscle/CKMM protein (Uniprot: P06732) | |
100 μg | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin), Protein Array | |
CKMM/3341 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
1158 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction