Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Creatinine Urinary Detection Kit

Catalog No. p-7229387
Encompass_Preferred
Click to view available options
Quantity:
2 x 96 Tests

Catalog No. EIACUN

Supplier: Invitrogen™ EIACUN

Only null left
Add to Cart
Edge
Add to Cart

Includes

96-well plates, standard, other assay specific reagents, buffers and lot-specific technical data sheet.

Assay Urinary Creatinine levels in Human, Monkey, Dog, and Rat Urine.

The Creatinine Urinary Detection research-use-only kit is a colorimetric assay designed for the quantification and detection of creatinine in human, rat, dog and monkey urine samples. Mouse urine samples are not compatible with the use of this assay for the detection of glomerular filtration rate (creatinine clearance).

This complete, ready-to-use kit includes clear 96-well plate(s), creatinine standard, creatinine detection reagent, and other components to perform the assay. A 96-well microplate reader capable of reading optical density at 490 nm, preferably with correction between 570 and 590 nm, is required for use of this kit.

Performance characteristics
• Assay type: colorimetric detection kit
• Sample types: human, rat, dog, and monkey urine samples
• Sensitivity: 0.019 mg/dL
• Standard curve range: 0.3–20 mg/dL
• Reactivity: species independent

Background
Creatinine (2-amino-1-methyl-5H-imadazol-4-one) is a metabolite of phosphocreatine (p-creatine), a molecule used as a store for high-energy phosphate that can be utilized by tissues for the production of ATP. Creatine either comes from the diet or synthesized from the amino acids arginine, glycine, and methionine. This occurs in the kidneys and liver, although other organ systems may be involved and species-specific differences may exist. Creatine and p-creatine are converted non-enzymatically to the metabolite creatinine, which diffuses into the blood and is excreted by the kidneys. In vivo, this conversion appears to be irreversible and in vitro it is favored by higher temperatures and lower pH. Creatinine forms spontaneously from p-creatine. This assay should to measure Creatinine from a wide range of sources.

Assay principle
The Creatinine Urinary Detection kit is designed to quantitatively measure creatinine present in urine samples. A creatinine standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate. The color generating reaction is initiated with the Creatinine Detection Reagent, which is pipetted into each well. After a short incubation the intensity of the generated color is detected in a microtiter plate reader capable of measuring 490 nm wavelength. The Jaffe reaction used in this kit has been modified to read creatinine levels in urine.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 96-well plate(s), standard, other assay specific reagents, buffers, and lot-specific technical data sheet.

Store at –20°C.
Detection Method Colorimetric
Immunoassay Kit Format Colorimetric Detection Kit
Product Type Creatinine Urinary Detection Kit
Sample Type Urine
For Use With (Equipment) Microplate Reader
Gene ID (Entrez) Non-Gene
Gene Symbol Non-Gene
Quantity 2 x 96 Tests
Shipping Condition Approved for shipment on Wet or Dry Ice
Target Kit Named Creatinine Urinary
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.