Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CREB, p Ser133 Antibody (SR1800), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | CREB |
---|---|
Clone | SR1800 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:40, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:40 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
CREB Monoclonal antibody specifically detects CREB in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
CREB | |
Western Blot 1:500-1:2000, Flow Cytometry 1:40, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:40 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein | |
A synthesized peptide derived from human phospho-Creb [Ser133] (Uniprot #: P16220) | |
Primary | |
Store at -20 degrees C. Avoid freeze/thaw cycles. |
SR1800 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Chromatin Research, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1385 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title