Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CREB1, Rabbit anti-Human, Clone: 4D11, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB18871
Specifications
CREB1 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.4 (50% glycerol, 0.1% BSA and 0.02% sodium azide). | |
CREB/MGC9284 | |
Rabbit | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry, Western Blot | |
4D11 | |
Flow Cytometry Western Blot The optimal working dilution should be determined by the end user. | |
cAMP responsive element binding protein 1 | |
CREB1 | |
A synthetic phosphopeptide corresponding to residues surrounding S133 of human CREB1. | |
RUO | |
1385 | |
Store at -20°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction