Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CREB3L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144901
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144901 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144901 Supplier Invitrogen™ Supplier No. PA5144901
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATTPLLGLS PLSRLPIPHQ APGEMTQLPV IKAEPLEVNQ FLKVTPEDLV QMPPTP Antigen sequence identity: Mouse 89%; Rat 89%.

Transcription factor that acts during endoplasmic reticulum stress by activating unfolded protein response target genes. Specifically involved in ER-stress response in astrocytes in the central nervous system. May play a role in gliosis. In vitro, binds to box-B element, cAMP response element (CRE) and CRE-like sequences, and activates transcription through box-B element but not through CRE.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB3L1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CREB3L1
Gene Accession No. Q96BA8
Gene Alias BBF-2 homolog; cAMP responsive element binding protein 3 like 1; cAMP responsive element binding protein 3-like 1; cAMP-responsive element-binding protein 3-like protein 1; CREB3L1; Cyclic AMP-responsive element-binding protein 3-like protein 1; OA-N; Oasis; Oasis N-terminal fragment; old astrocyte specifically induced substance; old astrocyte specifically-induced substance; Processed cyclic AMP-responsive element-binding protein 3-like protein 1; PSEC0238
Gene Symbols CREB3L1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 153-208. Recombinant protein control fragment (Product #RP-110090).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90993
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.