Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CREB3L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518417
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518417 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518417 Supplier Invitrogen™ Supplier No. PA518417
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Transcription factor that acts during endoplasmic reticulum stress by activating unfolded protein response target genes. Specifically involved in ER-stress response in astrocytes in the central nervous system. May play a role in gliosis. In vitro, binds to box-B element, cAMP response element (CRE) and CRE-like sequences, and activates transcription through box-B element but not through CRE.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB3L1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CREB3L1
Gene Accession No. Q96BA8
Gene Alias BBF-2 homolog; cAMP responsive element binding protein 3 like 1; cAMP responsive element binding protein 3-like 1; cAMP-responsive element-binding protein 3-like protein 1; CREB3L1; Cyclic AMP-responsive element-binding protein 3-like protein 1; OA-N; Oasis; Oasis N-terminal fragment; old astrocyte specifically induced substance; old astrocyte specifically-induced substance; Processed cyclic AMP-responsive element-binding protein 3-like protein 1; PSEC0238
Gene Symbols CREB3L1
Host Species Goat
Immunogen Synthetic peptide sequence (DFSHSKEWFHDRD) corresponding to the C-terminus amino acids of CREB3L1 (aa 497-510).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90993
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.