Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CREB3L3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CREB3L3 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CREB3L3 Polyclonal specifically detects CREB3L3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CREB3L3 | |
Polyclonal | |
Rabbit | |
Human | |
Q68CJ9 | |
84699 | |
This antibody was developed against a recombinant protein corresponding to amino acids: DAVPGSEAPGPRPEADTTREESPGSPGADWGFQDTANLTNSTEELDNATLVLRNATEGLGQVALLDWVAPGPSTGSGRAGLE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
cAMP responsive element binding protein 3-like 3, cAMP-responsive element-binding protein 3-like protein 3, CREB/ATF family transcription factor, CREB-H, CREBHMGC126557, cyclic AMP-responsive element-binding protein 3-like protein 3, MGC126553, Transcription factor CREB-H | |
CREB3L3 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title