Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CREB3L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563270
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563270 Supplier Invitrogen™ Supplier No. PA563270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DAVPGSEAPG PRPEADTTRE ESPGSPGADW GFQDTANLTN STEELDNATL VLRNATEGLG QVALLDWVAP GPSTGSGRAG LE Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 49%.

CREB3L3 is a liver-specific transcription factor and member of the CREB/ATF transcription factor family (Omori et al., 2001 [PubMed 11353085]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB3L3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CREB3L3
Gene Accession No. Q68CJ9
Gene Alias BC010786; cAMP responsive element binding protein 3 like 3; cAMP responsive element binding protein 3-like 3; cAMP-responsive element-binding protein 3-like protein 3; CREB/ATF family transcription factor; CREB3L3; CREBH; CREB-H; Cyclic AMP-responsive element-binding protein 3-like protein 3; D10Bur1e; HYST1481; Processed cyclic AMP-responsive element-binding protein 3-like protein 3; Transcription factor CREB-H
Gene Symbols CREB3L3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CREB3L3. Recombinant protein control fragment (Product #RP-103249).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84699
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.