Learn More
Invitrogen™ CREB3L4 Monoclonal Antibody (1165CT16.1.1)

Description
CREB3L4 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | CREB3L4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1165CT16.1.1 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CREB3L4 |
| Gene Accession No. | Q5UEM7, Q8TEY5 |
| Gene Alias | 1700012K17Rik; 5330432F22Rik; acre1; AIBZIP; Androgen-induced basic leucine zipper protein; androgen-induced bZIP protein; ATCE1; Attaching to CRE-like 1; AV040530; AV258827; cAMP responsive element binding protein 1; cAMP responsive element binding protein 3 like 4; cAMP responsive element binding protein 3-like 4; cAMP-responsive element-binding protein 3-like protein 4; cAMP-responsive element-binding protein 4; CREB3; Creb3l4; CREB4; CREB-4; Cyclic AMP-responsive element-binding protein 3-like protein 4; cyclic AMP-responsive element-binding protein 4; hJAL; JAL; mJAL; OTTHUMP00000035266; Processed cyclic AMP-responsive element-binding protein 3-like protein 4; RP11-422P24.8; Tisp40; transcript induced in spermiogenesis protein 40 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.