Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CREB3L4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518028
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518028 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518028 Supplier Invitrogen™ Supplier No. PA518028
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

cAMP response element-binding (CREB) proteins are a familyof mammalian transcription activators. For further backgroundinformation on CREB proteins, see CREB1 (MIM 123810).[supplied byOMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB3L4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CREB3L4
Gene Accession No. Q8TEY5
Gene Alias 1700012K17Rik; 5330432F22Rik; acre1; AIBZIP; Androgen-induced basic leucine zipper protein; androgen-induced bZIP protein; ATCE1; Attaching to CRE-like 1; AV040530; AV258827; cAMP responsive element binding protein 1; cAMP responsive element binding protein 3 like 4; cAMP responsive element binding protein 3-like 4; cAMP-responsive element-binding protein 3-like protein 4; cAMP-responsive element-binding protein 4; CREB3; Creb3l4; CREB4; CREB-4; Cyclic AMP-responsive element-binding protein 3-like protein 4; cyclic AMP-responsive element-binding protein 4; hJAL; JAL; mJAL; OTTHUMP00000035266; Processed cyclic AMP-responsive element-binding protein 3-like protein 4; RP11-422P24.8; Tisp40; transcript induced in spermiogenesis protein 40
Gene Symbols CREB3L4
Host Species Goat
Immunogen Synthetic peptide sequence (PSGRIRSVLHADEM) corresponding to the C-terminus amino acids of CREB3L4 (aa 382-395).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148327
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.