Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CREB3L4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558254
Description
Immunogen sequence: ELERHNISLV AQLRQLQTLI AQTSNKAAQT STCVLILLFS LALIILPSFS PFQSRPEAGS EDYQPHGVTS RNILTHKDVT ENLETQVVES RLREPPGAKD ANGSTR Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 71%.
cAMP response element-binding (CREB) proteins are a familyof mammalian transcription activators. For further backgroundinformation on CREB proteins, see CREB1 (MIM 123810).[supplied byOMIM].
Specifications
| CREB3L4 | |
| Polyclonal | |
| Unconjugated | |
| CREB3L4 | |
| 1700012K17Rik; 5330432F22Rik; acre1; AIBZIP; Androgen-induced basic leucine zipper protein; androgen-induced bZIP protein; ATCE1; Attaching to CRE-like 1; AV040530; AV258827; cAMP responsive element binding protein 1; cAMP responsive element binding protein 3 like 4; cAMP responsive element binding protein 3-like 4; cAMP-responsive element-binding protein 3-like protein 4; cAMP-responsive element-binding protein 4; CREB3; Creb3l4; CREB4; CREB-4; Cyclic AMP-responsive element-binding protein 3-like protein 4; cyclic AMP-responsive element-binding protein 4; hJAL; JAL; mJAL; OTTHUMP00000035266; Processed cyclic AMP-responsive element-binding protein 3-like protein 4; RP11-422P24.8; Tisp40; transcript induced in spermiogenesis protein 40 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 148327 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TEY5 | |
| CREB3L4 | |
| Recombinant protein corresponding to Human CREB3L4. Recombinant protein control fragment (Product #RP-89174). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction