Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CREB3L4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103498
Description
Antibody detects endogenous levels of total CREB3L4.
cAMP response element-binding (CREB) proteins are a familyof mammalian transcription activators. For further backgroundinformation on CREB proteins, see CREB1 (MIM 123810).[supplied byOMIM].
Specifications
CREB3L4 | |
Polyclonal | |
Unconjugated | |
CREB3L4 | |
1700012K17Rik; 5330432F22Rik; acre1; AIBZIP; Androgen-induced basic leucine zipper protein; androgen-induced bZIP protein; ATCE1; Attaching to CRE-like 1; AV040530; AV258827; cAMP responsive element binding protein 1; cAMP responsive element binding protein 3 like 4; cAMP responsive element binding protein 3-like 4; cAMP-responsive element-binding protein 3-like protein 4; cAMP-responsive element-binding protein 4; CREB3; Creb3l4; CREB4; CREB-4; Cyclic AMP-responsive element-binding protein 3-like protein 4; cyclic AMP-responsive element-binding protein 4; hJAL; JAL; mJAL; OTTHUMP00000035266; Processed cyclic AMP-responsive element-binding protein 3-like protein 4; RP11-422P24.8; Tisp40; transcript induced in spermiogenesis protein 40 | |
Rabbit | |
Affinity chromatography | |
RUO | |
148327, 310616, 78284 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5UEM7, Q8TEY5, Q9D2A5 | |
CREB3L4 | |
A synthesized peptide derived from human CREB3L4(Accession Q8TEY5), corresponding to amino acid residues R236-Q286. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction