Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CREB3L4 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | CREB3L4 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8891
![]()
|
Proteintech Group Inc
136301AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
cAMP response element-binding (CREB) proteins are a familyof mammalian transcription activators. For further backgroundinformation on CREB proteins, see CREB1 (MIM 123810).[supplied byOMIM].Specifications
| CREB3L4 | |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8TEY5 | |
| 148327 | |
| CREB3L4 Fusion Protein Ag4548 | |
| Primary | |
| -20°C | |
| CREB3L4 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| AIBZIP, ATCE1, Attaching to CRE like 1, CREB 4, CREB3, CREB3L4, CREB4, hJAL, JAL, Tisp40 | |
| CREB3L4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title