Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CREB5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107107
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107107 Supplier Invitrogen™ Supplier No. PA5107107
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 93%; Mouse: 93%; Rabbit: 93%; Rat: 93%.

The product of this gene belongs to the CRE (cAMP response element)-binding protein family. Members of this family contain zinc-finger and bZIP DNA-binding domains. The encoded protein specifically binds to CRE as a homodimer or a heterodimer with c-Jun or CRE-BP1, and functions as a CRE-dependent trans-activator. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene Creb5
Gene Accession No. Q8K1L0
Gene Alias cAMP response element-binding protein CRE-BPa; cAMP responsive element binding protein 5; cAMP-response element-binding protein A; cAMP-responsive element-binding protein 5; CREB5; CREB-5; CREBPA; CRE-BPa; Cyclic AMP-responsive element-binding protein 5; D430026C09Rik; RGD1566107
Gene Symbols Creb5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of Mouse Creb5 (aa 151-200).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231991
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.