Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CREB5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566406
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566406 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566406 Supplier Invitrogen™ Supplier No. PA566406
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSPPASPVPAC SQQQVIQHNT ITTSSSVSEV Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

The product of this gene belongs to the CRE (cAMP response element)-binding protein family. Members of this family contain zinc-finger and bZIP DNA-binding domains. The encoded protein specifically binds to CRE as a homodimer or a heterodimer with c-Jun or CRE-BP1, and functions as a CRE-dependent trans-activator. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Creb5
Gene Accession No. Q02930
Gene Alias cAMP response element-binding protein CRE-BPa; cAMP responsive element binding protein 5; cAMP-response element-binding protein A; cAMP-responsive element-binding protein 5; CREB5; CREB-5; CREBPA; CRE-BPa; Cyclic AMP-responsive element-binding protein 5; D430026C09Rik; RGD1566107
Gene Symbols Creb5
Host Species Rabbit
Immunogen Recombinant Human CREB5. Recombinant protein control fragment (Product #RP-105197).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9586
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.