Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CREB5 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS14053R

Description
The product of this gene belongs to the CRE (cAMP response element)-binding protein family. Members of this family contain zinc-finger and bZIP DNA-binding domains. The encoded protein specifically binds to CRE as a homodimer or a heterodimer with c-Jun or CRE-BP1, and functions as a CRE-dependent trans-activator. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
CREB5 | |
Polyclonal | |
Unconjugated | |
Creb5 | |
cAMP response element-binding protein CRE-BPa; cAMP responsive element binding protein 5; cAMP-response element-binding protein A; cAMP-responsive element-binding protein 5; Creb5; CREB-5; CREBPA; CRE-BPa; Cyclic AMP-responsive element-binding protein 5; D430026C09Rik; RGD1566107 | |
Rabbit | |
Protein A | |
RUO | |
9586 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q02930 | |
Creb5 | |
KLH conjugated synthetic peptide derived from human CREB5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction