Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CREBRF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568552
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568552 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568552 Supplier Invitrogen™ Supplier No. PA568552
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Acts as a negative regulator of the endoplasmic reticulum stress response or unfolded protein response (UPR). Represses the transcriptional activity of CREB3 during the UPR. Recruits CREB3 into nuclear foci.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREBRF
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CREBRF
Gene Accession No. Q8IUR6
Gene Alias A930001N09Rik; adult retina protein; C20H5orf41; C4H5orf41; C5orf41; CREB3 regulatory factor; CREBRF; gb:bc142922; LRF; Luman recruitment factor; luman/CREB3 recruitment factor; luman-recruiting factor; RGD1310862; UPF0474 protein C5orf41; UPF0474 protein C5orf41 homolog
Gene Symbols CREBRF
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human CREBRF.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153222
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.