Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CREBZF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530787
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530787 Supplier Invitrogen™ Supplier No. PA530787
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: mouse brain. Predicted reactivity: Mouse (88%), Rat (90%), Rhesus Monkey (97%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Strongly activates transcription when bound to HCFC1. Suppresses the expression of HSV proteins in cells infected with the virus in a HCFC1-dependent manner. Also suppresses the HCFC1-dependent transcriptional activation by CREB3 and reduces the amount of CREB3 in the cell. Able to down-regulate expression of some cellular genes in CREBZF-expressing cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREBZF
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene CREBZF
Gene Accession No. Q91ZR3, Q9NS37
Gene Alias 1110034C16Rik; 6330417B10Rik; CREB/ATF bZIP transcription factor; Crebzf; HCF-binding transcription factor Zhangfei; host cell factor-binding transcription factor Zhangfei; LAZip; LAZipII; RGD1565584; SHP-interacting leucine zipper protein; SMILE; Tyrosine kinase-associated leucine zipper protein LAZip; tyrosine kinase-associated leucine zipper protein LAZipII; ZF; Zhangfei
Gene Symbols CREBZF
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CREBZF. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233490, 58487
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.