Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREBZF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554439
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554439 Supplier Thermo Scientific Supplier No. PA554439
Add to Cart
Add to Cart

CREBZF Polyclonal Antibody

CREBZF Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen CREBZF
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CREBZF
Gene Accession No. Q9NS37
Gene Alias 1110034C16Rik; 6330417B10Rik; CREB/ATF bZIP transcription factor; Crebzf; HCF-binding transcription factor Zhangfei; host cell factor-binding transcription factor Zhangfei; LAZip; LAZipII; RGD1565584; SHP-interacting leucine zipper protein; SMILE; Tyrosine kinase-associated leucine zipper protein LAZip; tyrosine kinase-associated leucine zipper protein LAZipII; ZF; Zhangfei
Gene Symbols CREBZF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CREBZF. Recombinant protein control fragment (Product #RP-91635).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58487
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.