Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CREG Antibody (1C6), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242689B
Description
CREG Monoclonal specifically detects CREG in Human samples. It is validated for Western Blot, ELISA.Specifications
CREG | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CREG1 | |
Recombinant human CREG1 (32-220aa) purified from E. coli | |
100 μL | |
Cancer, Cell Cycle and Replication | |
8804 | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, ELISA | |
1C6 | |
Western Blot, ELISA | |
cellular repressor of E1A-stimulated genes, cellular repressor of E1A-stimulated genes 1CREG, protein CREG1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction