Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CREG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566012
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566012 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566012 Supplier Invitrogen™ Supplier No. PA566012
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YATLTMTLAQT NFCKKHGFDP QSPLCVHIML SGTVTKVNET EMDIAKHSLF IRHPEMKTWP SSHNWFFAKL NITNIWLLD Highest antigen sequence identity to the following orthologs - mouse 78%, rat 76%.

The adenovirus E1A protein both activates and represses gene expression to promote cellular proliferation and inhibit differentiation. CREG1 antagonizes transcriptional activation and cellular transformation by E1A. This protein shares limited sequence similarity with E1A and binds both the general transcription factor TBP and the tumor suppressor pRb in vitro.The adenovirus E1A protein both activates and represses gene expression to promote cellular proliferation and inhibit differentiation. The protein encoded by this gene antagonizes transcriptional activation and cellular transformation by E1A. This protein shares limited sequence similarity with E1A and binds both the general transcription factor TBP and the tumor suppressor pRb in vitro. This gene may contribute to the transcriptional control of cell growth and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREG1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CREG1
Gene Accession No. O75629
Gene Alias AA755314; cellular repressor of E1A stimulated genes 1; cellular repressor of E1A-stimulated genes 1; CREG; CREG1; Protein CREG1; UNQ727/PRO1409
Gene Symbols CREG1
Host Species Rabbit
Immunogen Recombinant Human CREG1. Recombinant protein control fragment (Product #RP-106696).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8804
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.