Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CREG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558302
Description
Immunogen sequence: RCVQLTLTGQ MIAVSPEEVE FAKQAMFSRH PGMRKWPRQY EWFFMKMRIE HIWLQKWYGG ASSISREEYF KAVP Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 61%.

Specifications
CREG2 | |
Polyclonal | |
Unconjugated | |
CREG2 | |
A830098L22Rik; cellular repressor of E1A stimulated genes 2; cellular repressor of E1A-stimulated genes 2; cellular repressor of E1A-stimulated genes 2, pseudogene 1; CREG2; Creg2-ps1; Protein CREG2; RGD1564056 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
200407 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IUH2 | |
CREG2 | |
Recombinant protein corresponding to Human CREG2. Recombinant protein control fragment (Product #RP-96204). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction