Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRELD1 Antibody (116), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290218PCP
Description
CRELD1 Monoclonal antibody specifically detects CRELD1 in Human samples. It is validated for ELISASpecifications
CRELD1 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
78987 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
116 | |
ELISA | |
atrioventricular septal defect 2, AVSD2, CIRRIN, cysteine-rich with EGF-like domain protein 1, cysteine-rich with EGF-like domains 1, DKFZp566D213 | |
Rabbit immunized with purified, recombinant Human CRELD1 (Accession#: NP_001070883.1, Met1-Glu362) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction