Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRELD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547581
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547581 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547581 Supplier Invitrogen™ Supplier No. PA547581
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, approximately 15% cross-reactivity with recombinant human CRELD1 is observed and less than 5% cross-reactivity with recombinant mouse CRELD2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Epidermal growth factor (EGF; MIM 131530)-like repeats are a class of cysteine-rich domains that mediate interactions between proteins of diverse function. EGF domains are found in proteins that are either completely secreted or have transmembrane regions that tether the protein to the cell surface. CRELD1 is the founding member of a family of matricellular proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRELD1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Creld1
Gene Accession No. Q91XD7
Gene Alias AI843811; AVSD2; CIRRIN; CRELD1; cysteine rich with EGF like domains 1; cysteine rich with EGF-like domains 1; cysteine-rich with EGF-like domain protein 1; cysteine-rich with EGF-like domains 1; DKFZP566D213; i11E7 protein; protein disulfide isomerase CRELD1; UNQ188/PRO214
Gene Symbols Creld1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CRELD1 Gln30-Glu362.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171508
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.