Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRELD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555468
missing translation for 'changeView'
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. PIPA555468 Supplier Invitrogen™ missing translation for 'supplierNo' PA555468
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAFPILTDLT PETTRRWKLG SHPHSTYVKM KMQRDEATFP GLYGKQVAKL GSQSRQSDRG TRL Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 30%.

Epidermal growth factor (EGF; MIM 131530)-like repeats are a class of cysteine-rich domains that mediate interactions between proteins of diverse function. EGF domains are found in proteins that are either completely secreted or have transmembrane regions that tether the protein to the cell surface. CRELD1 is the founding member of a family of matricellular proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRELD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Creld1
Gene Accession No. Q96HD1
Gene Alias AI843811; AVSD2; CIRRIN; CRELD1; cysteine rich with EGF like domains 1; cysteine rich with EGF-like domains 1; cysteine-rich with EGF-like domain protein 1; cysteine-rich with EGF-like domains 1; DKFZP566D213; i11E7 protein; protein disulfide isomerase CRELD1; UNQ188/PRO214
Gene Symbols Creld1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRELD1. Recombinant protein control fragment (Product #RP-91008).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78987
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.