Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRELD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590280
Description
Immunogen sequence: RGLVDSFNKG LERTIRDNFG GGNTAWEEEN LSKYKDSETR LVEVLEGVCS KSDFECHRLL ELSEELVESW WFHKQQEAPD LFQWLCSDSL KLCCPAGTFG P; Positive Samples: Mouse lung; Cellular Location: Membrane, Multi-pass membrane protein.
Epidermal growth factor (EGF; MIM 131530)-like repeats are a class of cysteine-rich domains that mediate interactions between proteins of diverse function. EGF domains are found in proteins that are either completely secreted or have transmembrane regions that tether the protein to the cell surface. CRELD1 is the founding member of a family of matricellular proteins.
Specifications
CRELD1 | |
Polyclonal | |
Unconjugated | |
Creld1 | |
AI843811; AVSD2; CIRRIN; CRELD1; cysteine rich with EGF like domains 1; cysteine rich with EGF-like domains 1; cysteine-rich with EGF-like domain protein 1; cysteine-rich with EGF-like domains 1; DKFZP566D213; i11E7 protein; protein disulfide isomerase CRELD1; UNQ188/PRO214 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
171508, 312638, 78987 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.08 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q4V7F2, Q91XD7, Q96HD1 | |
Creld1 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human CRELD1 (NP_0563282). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction