Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRELD1 Recombinant Rabbit Monoclonal Antibody (116)
GREENER_CHOICE

Catalog No. PIMA530274
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530274 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530274 Supplier Invitrogen™ Supplier No. MA530274
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human CRELD1.

Epidermal growth factor (EGF; MIM 131530)-like repeats are a class of cysteine-rich domains that mediate interactions between proteins of diverse function. EGF domains are found in proteins that are either completely secreted or have transmembrane regions that tether the protein to the cell surface. CRELD1 is the founding member of a family of matricellular proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRELD1
Applications ELISA
Classification Recombinant Monoclonal
Clone 116
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Creld1
Gene Accession No. Q96HD1
Gene Alias AI843811; AVSD2; CIRRIN; CRELD1; cysteine rich with EGF like domains 1; cysteine rich with EGF-like domains 1; cysteine-rich with EGF-like domain protein 1; cysteine-rich with EGF-like domains 1; DKFZP566D213; i11E7 protein; protein disulfide isomerase CRELD1; UNQ188/PRO214
Gene Symbols Creld1
Host Species Rabbit
Immunogen Recombinant Human CRELD1 protein (Met1-Glu362).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78987
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.