Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREM Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986357
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6357 Supplier Bioss Supplier No. BS5135R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a bZIP transcription factor that binds to the cAMP responsive element found in many viral and cellular promoters. It is an important component of cAMP-mediated signal transduction during the spermatogenetic cycle, as well as other complex processes. Alternative promoter and translation initiation site usage allows this gene to exert spatial and temporal specificity to cAMP responsiveness. Multiple alternatively spliced transcript variants encoding several different isoforms have been found for this gene, with some of them functioning as activators and some as repressors of transcription.

Specifications

Antigen CREM
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CREM
Gene Accession No. Q03060
Gene Alias cAMP response element modulator; cAMP responsive element moderator; cAMP responsive element modulator; cAMP-responsive element modulator; CRE modulator; CREM; CREM 2alpha-b protein; CREM 2beta-a protein; CREM-2; hCREM-2; ICER; ICERI; Inducible cAMP early repressor; inducible cAMP early repressor ICER; inducible cyclic AMP early repressor; MGC111110; MGC17881; MGC41893; OTTHUMP00000019443
Gene Symbols CREM
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CREM.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1390
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.