Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Cresol Red sodium salt

Catalog No. p-7025719
Click to view available options
Quantity:
5 g
25 g
100 g

Transition interval (acid): pH 0.2 (red) to pH 1.8 (yellow); (alkaline) pH 7.2 (yellow) to pH 8.8 (red) | CAS: 62625-29-0 | C21H17NaO5S

Cresol Red sodium salt acts as a constituent of polymerase chain reaction reagent systems. It is useful as a pH indicator with color changes with pH 0.2 - pH 1.8 and pH 7.1 - 8.8. Further, it plays an important role in molecular biology reactions in place of other loading dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cresol Red sodium salt acts as a constituent of polymerase chain reaction reagent systems. It is useful as a pH indicator with color changes with pH 0.2 - pH 1.8 and pH 7.1 - 8.8. Further, it plays an important role in molecular biology reactions in place of other loading dyes.

Solubility
Soluble in water, dilute alcohols, acids, alkalies and ethanol.

Notes
Transition interval (acid): pH 0.2 (red) to pH 1.8 (yellow); (alkaline) pH 7.2 (yellow) to pH 8.8 (red)Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-29-0
MDL Number MFCD00001618

Specifications

Melting Point ∼250°C (decomposition)
Molecular Formula C21H17NaO5S
Solubility Information Soluble in water,dilute alcohols,acids,alkalies and ethanol.
Formula Weight 404.42
Quantity 5 g
Chemical Name or Material Cresol Red sodium salt
EINECSNumber 263-654-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.