Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CREST Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100094
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100094 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100094 Supplier Invitrogen™ Supplier No. PA5100094
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CREST.

Synovial sarcomas occur most frequently in the extremities around large joints. More than 90% of cases have a recurrent and specific chromosomal translocation, t(X;18)(p11.2;q11.2), in which the 5-prime end of the SS18 gene (MIM 600192) is fused in-frame to the 3-prime end of the SSX1 (MIM 312820), SSX2 (MIM 300192), or SSX4 (MIM 300326) gene. The SS18L1 gene is homologous to SS18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREST
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SS18L1
Gene Accession No. O75177, Q8BW22, Q91XJ0
Gene Alias A230053O16Rik; Calcium-responsive transactivator; calcium-responsive transcription coactivator; CREST; KIAA0693; LP2261; Ss18l1; SS18L1, nBAF chromatin remodeling complex subunit; SS18-like protein 1; synovial sarcoma translocation gene on chromosome 18-like 1; SYT homolog 1
Gene Symbols SS18L1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SS18L1(Accession O75177), corresponding to amino acid residues S258-S308.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192352, 26039, 269397
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.