Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cresyl Violet acetate, pure, certified

Catalog No. AC405760250
Click to view available options
Quantity:
2.5 g
10 g
25 g
50 g
Packaging:
Glass bottle

CAS: 10510-54-0 | C18H15N3O3 | 321.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10510-54-0
Molecular Formula C18H15N3O3
Molecular Weight (g/mol) 321.34
MDL Number MFCD00013151,MFCD00013151
InChI Key XKOCOMKJPWEOHX-UHFFFAOYSA-M
PubChem CID 44134641
SMILES CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3

Specifications

Absorption Ratio 0.94 to 1.2
Dye Content 65% min.
Infrared Spectrum Authentic
IUPAC Name 8,12-diamino-10λ⁴-oxa-5-azatetraphen-10-ylium acetate
Lambda Max 596 to 599nm
Formula Weight 321.34
Grade Pure
Packaging Glass bottle
Color Black to Green
Quantity 25 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Cresyl Violet acetate
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.