Learn More
CRF Monoclonal Antibody (5D5), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA183085
Description
Predicted to react with rat based on sequence homology. This product requires protein digestion pre-treatment of paraffin sections using trypsin or pronase. Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. A suggested positive control for this product is human hypothalamus. Mouse anti Human corticotrophin-releasing factor antibody, clone 5D5 recognizes human corticotropin-releasing factor (CRF), a secreted hormone which regulates the release of corticotropin from the pituitary.
Corticotropin Releasing Factor (CRF), a 41 amino acids neuropeptide, belongs to a family of structurally related peptides including urocortin, sauvagine and urotensin I. CRF is critical in the regulation of the pituitary-adrenal gland axis, and in endocrine, autonomic and behavioral responses to stress. CRF is widely distributed in the brain and peripheral nervous system with the highest levels in the hypothalamic paraventricular nucleus.
Specifications
CRF | |
Monoclonal | |
1 mg/mL | |
PBS with <0.1% sodium azide | |
P06850 | |
CRH | |
Synthetic human corticotropin factor | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin) | |
5D5 | |
Unconjugated | |
CRH | |
Corticoliberin; corticotropin releasing factor; corticotropin releasing hormone; corticotropin releasing hormone receptor 2; corticotropin-releasing factor; Corticotropin-releasing factor receptor 2; corticotropin-releasing hormone; Corticotropin-releasing hormone receptor 2; CRF; CRF2; CRF2 receptor, beta isoform; CRF2R; CRFR2; CRF-R-2; CRF-RB; CRH; CRH receptor 2 variant B; CRH1; CRH2R; CRHR2; CRH-R2; CRH-R-2; Gm1347; HM-CRF | |
Mouse | |
Protein G | |
RUO | |
1392 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.